Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CD3E
Entrez Gene ID
916
Gene Name
CD3e molecule
Gene Aliases
IMD18, T3E, TCRE
Chromosome Location
Chr.11: 118304580 - 118316175 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000733.3 NP_000724.1 1 - 2 201 88
GenBank mRNA AK292612.1 - 1 - 2 50 88
AK301255.1 - 1 - 2 51 88
AK311641.1 - 1 - 2 51 88
BI910359.1 - 1 - 2 236 88

View Full Product Details