The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020639.2 | NP_065690.2 | 2 - 3 | 521 | 67 |
GenBank mRNA | AB047783.1 | - | 2 - 3 | 497 | 67 |
AJ278016.1 | - | 2 - 3 | 521 | 67 | |
AK027424.1 | - | 2 - 3 | 538 | 67 | |
AK302592.1 | - | 2 - 3 | 332 | 67 | |
AK316412.1 | - | 2 - 3 | 484 | 67 | |
BC035755.1 | - | 2 - 3 | 475 | 67 | |
BC110617.1 | - | 2 - 3 | 494 | 67 | |
BC110618.1 | - | 2 - 3 | 494 | 67 |