The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020639.2 | NP_065690.2 | 3 - 4 | 669 | 58 |
GenBank mRNA | AB047783.1 | - | 3 - 4 | 645 | 58 |
AJ278016.1 | - | 3 - 4 | 669 | 58 | |
AK302592.1 | - | 3 - 4 | 480 | 58 | |
AK316412.1 | - | 3 - 4 | 632 | 58 | |
BC035755.1 | - | 3 - 4 | 623 | 58 | |
BC110617.1 | - | 3 - 4 | 642 | 58 | |
BC110618.1 | - | 3 - 4 | 642 | 58 |