The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130105.1 | NP_001123577.1 | 7 - 8 | 1202 | 104 |
NM_005713.2 | NP_005704.1 | 6 - 7 | 1103 | 104 | |
NM_031361.2 | NP_112729.1 | 6 - 7 | 1103 | 104 | |
XM_006714513.2 | XP_006714576.1 | 6 - 7 | 1103 | 104 | |
XM_011543090.2 | XP_011541392.1 | 6 - 7 | 1103 | 104 | |
XM_011543091.2 | XP_011541393.1 | 6 - 7 | 1103 | 104 | |
XM_017008919.1 | XP_016864408.1 | 6 - 7 | 1103 | 104 | |
GenBank mRNA | AF136450.1 | - | 6 - 7 | 1089 | 104 |
AF232930.1 | - | 6 - 7 | 1071 | 104 | |
AK000604.1 | - | 2 - 3 | 123 | 104 | |
AK091851.1 | - | 7 - 8 | 993 | 104 | |
AK096854.1 | - | 7 - 8 | 1359 | 104 | |
AK292087.1 | - | 6 - 7 | 1103 | 104 | |
AY453385.1 | - | 6 - 7 | 681 | 104 | |
AY453386.1 | - | 6 - 7 | 681 | 104 | |
BC000102.2 | - | 6 - 7 | 1087 | 104 |