Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAF1
Entrez Gene ID
54623
Gene Name
PAF1 homolog, Paf1/RNA polymerase II complex component
Gene Aliases
F23149_1, PD2
Chromosome Location
Chr.19: 39385629 - 39391195 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256826.1 NP_001243755.1 5 - 6 759 67
NM_019088.3 NP_061961.2 6 - 7 789 67
NR_046384.1 5 - 6 667 67
GenBank mRNA AJ401156.1 - 6 - 7 614 67
AK098423.1 - 5 - 6 556 67
AK294625.1 - 5 - 6 589 67
BC000017.2 - 6 - 7 633 67
BC013402.1 - 6 - 7 705 67

View Full Product Details