The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127458.1 | NP_001120930.1 | 4 - 5 | 519 | 153 |
NM_001323561.1 | NP_001310490.1 | 4 - 5 | 527 | 153 | |
NM_001323562.1 | NP_001310491.1 | 4 - 5 | 534 | 153 | |
NM_001323563.1 | NP_001310492.1 | 4 - 5 | 902 | 153 | |
NM_001323564.1 | NP_001310493.1 | 4 - 5 | 528 | 153 | |
NM_019095.5 | NP_061968.1 | 4 - 5 | 840 | 153 | |
XM_005260738.4 | XP_005260795.1 | 4 - 5 | 504 | 153 | |
GenBank mRNA | AF241784.1 | - | 4 - 5 | 759 | 153 |
AL832419.1 | - | 4 - 5 | 587 | 153 | |
BM563835.1 | - | 4 - 5 | 522 | 153 | |
BP368984.1 | - | 4 - 5 | 519 | 153 | |
BU852955.1 | - | 5 - 6 | 547 | 153 | |
DQ386730.1 | - | 4 - 5 | 666 | 153 |