The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018225.2 | NP_060695.2 | 10 - 11 | 1399 | 74 |
XM_005251503.4 | XP_005251560.1 | 9 - 10 | 1235 | 74 | |
GenBank mRNA | AK001667.1 | - | 10 - 11 | 1329 | 74 |
AK225559.1 | - | 10 - 11 | 1354 | 74 | |
AK225587.1 | - | 10 - 11 | 1348 | 74 | |
AK304767.1 | - | 8 - 9 | 986 | 74 | |
BC002876.2 | - | 10 - 11 | 1345 | 74 | |
BC110654.1 | - | 10 - 11 | 1439 | 74 | |
EF011613.1 | - | 10 - 11 | 1345 | 74 |