The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018225.2 | NP_060695.2 | 2 - 3 | 349 | 69 |
GenBank mRNA | AK001667.1 | - | 2 - 3 | 279 | 69 |
AK225559.1 | - | 2 - 3 | 304 | 69 | |
AK225587.1 | - | 2 - 3 | 298 | 69 | |
BC002876.2 | - | 2 - 3 | 295 | 69 | |
BC110654.1 | - | 2 - 3 | 389 | 69 | |
EF011613.1 | - | 2 - 3 | 295 | 69 |