The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018225.2 | NP_060695.2 | 5 - 6 | 734 | 94 |
XM_005251503.4 | XP_005251560.1 | 4 - 5 | 570 | 94 | |
GenBank mRNA | AK001667.1 | - | 5 - 6 | 664 | 94 |
AK225559.1 | - | 5 - 6 | 689 | 94 | |
AK225587.1 | - | 5 - 6 | 683 | 94 | |
AK304767.1 | - | 3 - 4 | 321 | 94 | |
BC002876.2 | - | 5 - 6 | 680 | 94 | |
BC110654.1 | - | 5 - 6 | 774 | 94 | |
EF011613.1 | - | 5 - 6 | 680 | 94 |