The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018225.2 | NP_060695.2 | 6 - 7 | 852 | 112 |
XM_005251503.4 | XP_005251560.1 | 5 - 6 | 688 | 112 | |
GenBank mRNA | AK001667.1 | - | 6 - 7 | 782 | 112 |
AK225559.1 | - | 6 - 7 | 807 | 112 | |
AK225587.1 | - | 6 - 7 | 801 | 112 | |
AK304767.1 | - | 4 - 5 | 439 | 112 | |
BC002876.2 | - | 6 - 7 | 798 | 112 | |
BC110654.1 | - | 6 - 7 | 892 | 112 | |
EF011613.1 | - | 6 - 7 | 798 | 112 |