Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERCC4
Entrez Gene ID
2072
Gene Name
ERCC excision repair 4, endonuclease catalytic subunit
Gene Aliases
ERCC11, FANCQ, RAD1, XFEPS, XPF
Chromosome Location
Chr.16: 13920138 - 13952348 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005236.2 NP_005227.1 6 - 7 1105 116
XM_011522424.2 XP_011520726.1 7 - 8 1262 116
XM_017023043.1 XP_016878532.1 7 - 8 1219 116
GenBank mRNA AK308341.1 - 6 - 7 1071 116
U64315.1 - 6 - 7 1078 116

View Full Product Details