Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM68
Entrez Gene ID
137695
Gene Name
transmembrane protein 68
Gene Aliases
-
Chromosome Location
Chr.8: 55738744 - 55773407 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286657.1 NP_001273586.1 5 - 6 862 120
XM_005251150.1 XP_005251207.1 6 - 7 990 120
XM_011517454.1 XP_011515756.1 6 - 7 1014 120
XM_017013040.1 XP_016868529.1 5 - 6 574 120
GenBank mRNA AL832935.1 - 4 - 5 471 120

View Full Product Details