Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RUFY1
Entrez Gene ID
80230
Gene Name
RUN and FYVE domain containing 1
Gene Aliases
RABIP4, ZFYVE12
Chromosome Location
Chr.5: 179550551 - 179610026 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040451.2 NP_001035541.1 3 - 4 727 110
NM_001040452.2 NP_001035542.1 4 - 5 538 110
NM_025158.4 NP_079434.3 4 - 5 713 110
XM_006714921.3 XP_006714984.2 4 - 5 723 110
XM_017009891.1 XP_016865380.1 4 - 5 538 110
XM_017009892.1 XP_016865381.1 1 - 2 102 110
XM_017009893.1 XP_016865382.1 1 - 2 102 110
XM_017009894.1 XP_016865383.1 4 - 5 446 110
XM_017009895.1 XP_016865384.1 3 - 4 518 110
GenBank mRNA AB209507.1 - 4 - 5 438 110
AF312367.1 - 4 - 5 704 110
AF361055.2 - 4 - 5 686 110
AK075021.1 - 3 - 4 626 110
AK291926.1 - 3 - 4 727 110
BC032571.1 - 4 - 5 457 110
DA925727.1 - 4 - 5 499 110

View Full Product Details