Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RUFY1
Entrez Gene ID
80230
Gene Name
RUN and FYVE domain containing 1
Gene Aliases
RABIP4, ZFYVE12
Chromosome Location
Chr.5: 179550551 - 179610026 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040451.2 NP_001035541.1 4 - 5 847 81
NM_001040452.2 NP_001035542.1 5 - 6 658 81
NM_025158.4 NP_079434.3 5 - 6 833 81
XM_005265993.4 XP_005266050.1 2 - 3 204 81
XM_006714921.3 XP_006714984.2 5 - 6 843 81
XM_017009891.1 XP_016865380.1 5 - 6 658 81
XM_017009894.1 XP_016865383.1 5 - 6 566 81
XM_017009895.1 XP_016865384.1 4 - 5 638 81
GenBank mRNA AB209507.1 - 5 - 6 558 81
AF361055.2 - 5 - 6 806 81
AK075021.1 - 4 - 5 746 81
AK291926.1 - 4 - 5 847 81
BC032571.1 - 5 - 6 577 81

View Full Product Details