The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015131.2 | NP_055946.1 | 4 - 5 | 660 | 81 |
GenBank mRNA | AK000836.1 | - | 3 - 4 | 356 | 81 |
BC033206.1 | - | 3 - 4 | 365 | 81 | |
D26488.1 | - | 4 - 5 | 633 | 81 | |
D87716.1 | - | 2 - 3 | 139 | 81 |