Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WDR43
Entrez Gene ID
23160
Gene Name
WD repeat domain 43
Gene Aliases
NET12, UTP5
Chromosome Location
Chr.2: 28894643 - 28948220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015131.2 NP_055946.1 9 - 10 1234 131
GenBank mRNA AK000836.1 - 8 - 9 930 131
BC033206.1 - 8 - 9 939 131
D26488.1 - 9 - 10 1207 131
D87716.1 - 7 - 8 713 131

View Full Product Details