The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003886.2 | NP_003877.2 | 2 - 3 | 242 | 61 |
NM_139289.1 | NP_647450.1 | 2 - 3 | 215 | 61 | |
GenBank mRNA | AF072756.1 | - | 2 - 3 | 270 | 61 |
AK292463.1 | - | 2 - 3 | 236 | 61 | |
BC033007.1 | - | 2 - 3 | 239 | 61 | |
BC070150.1 | - | 2 - 3 | 234 | 61 | |
BC126250.1 | - | 2 - 3 | 241 | 61 | |
BC126252.1 | - | 2 - 3 | 241 | 61 | |
Y15195.1 | - | 2 - 3 | 242 | 61 |