The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003886.2 | NP_003877.2 | 3 - 4 | 297 | 65 |
NM_139289.1 | NP_647450.1 | 3 - 4 | 270 | 65 | |
GenBank mRNA | AF072756.1 | - | 3 - 4 | 325 | 65 |
AK292463.1 | - | 3 - 4 | 291 | 65 | |
BC033007.1 | - | 3 - 4 | 294 | 65 | |
BC070150.1 | - | 3 - 4 | 289 | 65 | |
BC126250.1 | - | 3 - 4 | 296 | 65 | |
BC126252.1 | - | 3 - 4 | 296 | 65 | |
Y15195.1 | - | 3 - 4 | 297 | 65 |