The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300743.1 | NP_001287672.1 | 8 - 9 | 1042 | 112 |
NM_020314.5 | NP_064710.4 | 8 - 9 | 1042 | 112 | |
XM_005255435.3 | XP_005255492.1 | 8 - 9 | 994 | 112 | |
XM_006721065.3 | XP_006721128.1 | 8 - 9 | 994 | 112 | |
XM_011545896.2 | XP_011544198.1 | 7 - 8 | 672 | 112 | |
GenBank mRNA | AK024693.1 | - | 4 - 5 | 312 | 112 |
AK095078.1 | - | 8 - 9 | 739 | 112 | |
AK290286.1 | - | 8 - 9 | 739 | 112 | |
AK294885.1 | - | 9 - 10 | 1097 | 112 | |
AL832159.1 | - | 8 - 9 | 738 | 112 | |
AL833428.1 | - | 8 - 9 | 731 | 112 | |
BC014900.2 | - | 7 - 8 | 710 | 112 | |
BC050464.1 | - | 8 - 9 | 782 | 112 | |
BC058845.1 | - | 8 - 9 | 741 | 112 | |
BX537867.1 | - | 8 - 9 | 754 | 112 |