The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317814.1 | NP_001304743.1 | 1 - 2 | 416 | 71 |
GenBank mRNA | BM552840.1 | - | 1 - 2 | 351 | 71 |