Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PLEKHG2
- Entrez Gene ID
-
64857
- Gene Name
- pleckstrin homology and RhoGEF domain containing G2
- Gene Aliases
-
ARHGEF42, CLG, LDAMD
- Chromosome Location
-
Chr.19: 39412582 - 39428415 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK026111.1 |
- |
17 - 18 |
1809 |
72 |
View Full Product Details