The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015120.4 | NP_055935.4 | 6 - 7 | 1454 | 111 |
GenBank mRNA | AJ417593.2 | - | 6 - 7 | 1454 | 111 |
AX751581.1 | - | 5 - 6 | 1328 | 111 |