The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287223.1 | NP_001274152.1 | 18 - 19 | 3090 | 82 |
NM_014139.2 | NP_054858.2 | 17 - 18 | 3259 | 82 | |
XM_011533321.2 | XP_011531623.1 | 14 - 15 | 2484 | 82 | |
XM_017005647.1 | XP_016861136.1 | 22 - 23 | 3711 | 82 | |
XM_017005648.1 | XP_016861137.1 | 19 - 20 | 3192 | 82 | |
XM_017005649.1 | XP_016861138.1 | 21 - 22 | 3589 | 82 | |
XM_017005650.1 | XP_016861139.1 | 18 - 19 | 3390 | 82 | |
XM_017005651.1 | XP_016861140.1 | 18 - 19 | 2916 | 82 | |
XM_017005652.1 | XP_016861141.1 | 20 - 21 | 3390 | 82 | |
XM_017005653.1 | XP_016861142.1 | 8 - 9 | 1868 | 82 | |
GenBank mRNA | AF109737.1 | - | 17 - 18 | 3259 | 82 |
AF150882.1 | - | 17 - 18 | 3259 | 82 | |
AF188679.1 | - | 18 - 19 | 3090 | 82 | |
AJ417790.1 | - | 17 - 18 | 3060 | 82 | |
AY686224.1 | - | 2 - 3 | 177 | 82 |