The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267594.1 | NP_001254523.1 | 16 - 17 | 2104 | 119 |
NM_001267595.1 | NP_001254524.1 | 16 - 17 | 2359 | 119 | |
NR_051991.1 | 17 - 18 | 2545 | 119 | ||
NR_051992.1 | 17 - 18 | 2558 | 119 | ||
XM_006719361.3 | XP_006719424.1 | 16 - 17 | 2239 | 119 | |
XM_006719362.2 | XP_006719425.1 | 17 - 18 | 2011 | 119 | |
XM_011538244.2 | XP_011536546.1 | 15 - 16 | 2240 | 119 | |
XM_011538245.2 | XP_011536547.1 | 15 - 16 | 2191 | 119 | |
XM_017019229.1 | XP_016874718.1 | 17 - 18 | 2377 | 119 | |
XM_017019230.1 | XP_016874719.1 | 16 - 17 | 2293 | 119 | |
XM_017019231.1 | XP_016874720.1 | 17 - 18 | 2277 | 119 | |
XM_017019232.1 | XP_016874721.1 | 17 - 18 | 2004 | 119 | |
XM_017019233.1 | XP_016874722.1 | 16 - 17 | 2287 | 119 | |
XM_017019234.1 | XP_016874723.1 | 16 - 17 | 2197 | 119 | |
XM_017019235.1 | XP_016874724.1 | 16 - 17 | 1930 | 119 | |
XM_017019236.1 | XP_016874725.1 | 15 - 16 | 2165 | 119 | |
XM_017019237.1 | XP_016874726.1 | 15 - 16 | 2065 | 119 | |
XM_017019238.1 | XP_016874727.1 | 17 - 18 | 2315 | 119 | |
XM_017019239.1 | XP_016874728.1 | 16 - 17 | 2355 | 119 | |
XM_017019240.1 | XP_016874729.1 | 16 - 17 | 2276 | 119 | |
GenBank mRNA | AK292060.1 | - | 16 - 17 | 1875 | 119 |
AK312874.1 | - | 16 - 17 | 2105 | 119 | |
BC045639.1 | - | 16 - 17 | 2175 | 119 | |
BX537920.1 | - | 17 - 18 | 2558 | 119 | |
BX640784.1 | - | 17 - 18 | 2389 | 119 |