The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286487.1 | NP_001273416.1 | 15 - 16 | 1599 | 78 |
NM_015497.4 | NP_056312.2 | 16 - 17 | 1660 | 78 | |
XM_005254287.1 | XP_005254344.1 | 16 - 17 | 1514 | 78 | |
XM_006720468.2 | XP_006720531.1 | 16 - 17 | 1519 | 78 | |
XM_011521443.2 | XP_011519745.1 | 15 - 16 | 1431 | 78 | |
XM_017022067.1 | XP_016877556.1 | 16 - 17 | 1516 | 78 | |
GenBank mRNA | AF132733.1 | - | 16 - 17 | 1516 | 78 |
AK074870.1 | - | 16 - 17 | 1509 | 78 | |
AK075403.1 | - | 16 - 17 | 1546 | 78 | |
AK289591.1 | - | 16 - 17 | 1509 | 78 | |
AK290150.1 | - | 16 - 17 | 1495 | 78 | |
AK290484.1 | - | 15 - 16 | 1443 | 78 | |
AK297816.1 | - | 16 - 17 | 1505 | 78 |