Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRCAP
Entrez Gene ID
10847
Gene Name
Snf2-related CREBBP activator protein
Gene Aliases
DOMO1, EAF1, FLHS, SWR1
Chromosome Location
Chr.16: 30699141 - 30740129 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006662.2 NP_006653.2 28 - 29 6685 83
GenBank mRNA AF143946.1 - 23 - 24 5733 83
AK128030.1 - 28 - 29 6685 83

View Full Product Details