Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FCHO1
Entrez Gene ID
23149
Gene Name
FCH domain only 1
Gene Aliases
-
Chromosome Location
Chr.19: 17747718 - 17788568 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161357.1 NP_001154829.1 22 - 23 2109 90
NM_001161358.1 NP_001154830.1 21 - 22 2019 90
NM_001161359.1 NP_001154831.1 20 - 21 1949 90
NM_015122.2 NP_055937.1 22 - 23 2113 90
XM_005259831.3 XP_005259888.1 21 - 22 2198 90
XM_006722701.3 XP_006722764.1 21 - 22 2117 90
XM_006722702.3 XP_006722765.1 21 - 22 2194 90
XM_011527824.2 XP_011526126.1 22 - 23 2109 90
XM_011527825.2 XP_011526127.1 22 - 23 2098 90
XM_011527826.2 XP_011526128.1 22 - 23 2081 90
XM_011527827.2 XP_011526129.1 19 - 20 1640 90
XM_011527828.2 XP_011526130.1 21 - 22 2012 90
XM_017026519.1 XP_016882008.1 22 - 23 2179 90
XM_017026520.1 XP_016882009.1 22 - 23 2162 90
XM_017026521.1 XP_016882010.1 21 - 22 2113 90
XM_017026522.1 XP_016882011.1 22 - 23 1992 90
XM_017026523.1 XP_016882012.1 22 - 23 2062 90
XM_017026524.1 XP_016882013.1 21 - 22 2077 90
XM_017026525.1 XP_016882014.1 21 - 22 1875 90
XM_017026526.1 XP_016882015.1 21 - 22 1833 90
XM_017026527.1 XP_016882016.1 20 - 21 1841 90
XM_017026528.1 XP_016882017.1 21 - 22 1903 90
GenBank mRNA AB006628.1 - 19 - 20 1883 90
AK291410.1 - 21 - 22 1975 90
AK303623.1 - 18 - 19 1811 90
AK309403.1 - 18 - 19 1862 90
AK316458.1 - 20 - 21 1920 90
BC028021.1 - 22 - 23 2054 90
BC041130.1 - 22 - 23 2063 90

View Full Product Details