The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018141.3 | NP_060611.2 | 2 - 3 | 162 | 96 |
XM_011514724.2 | XP_011513026.1 | 3 - 4 | 166 | 96 | |
GenBank mRNA | AF113220.1 | - | 2 - 3 | 122 | 96 |
AF277180.1 | - | 2 - 3 | 124 | 96 | |
AK001429.1 | - | 2 - 3 | 131 | 96 | |
AK098446.1 | - | 2 - 3 | 125 | 96 | |
AK316599.1 | - | 2 - 3 | 128 | 96 | |
BC012560.1 | - | 2 - 3 | 132 | 96 | |
BG776014.1 | - | 2 - 3 | 163 | 96 |