The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018141.3 | NP_060611.2 | 3 - 4 | 237 | 92 |
XM_005249206.2 | XP_005249263.1 | 3 - 4 | 246 | 92 | |
XM_011514724.2 | XP_011513026.1 | 4 - 5 | 241 | 92 | |
XM_011514725.2 | XP_011513027.1 | 4 - 5 | 233 | 92 | |
XM_017010996.1 | XP_016866485.1 | 3 - 4 | 238 | 92 | |
GenBank mRNA | AF113220.1 | - | 3 - 4 | 197 | 92 |
AF277180.1 | - | 3 - 4 | 199 | 92 | |
AK098446.1 | - | 3 - 4 | 200 | 92 | |
AK298219.1 | - | 3 - 4 | 220 | 92 | |
AK316599.1 | - | 3 - 4 | 203 | 92 | |
BC012560.1 | - | 3 - 4 | 207 | 92 | |
BG776014.1 | - | 3 - 4 | 238 | 92 |