The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024690.2 | NP_078966.2 | 65 - 66 | 41750 | 70 |
XM_017027486.1 | XP_016882975.1 | 59 - 60 | 40830 | 70 | |
XM_017027487.1 | XP_016882976.1 | 58 - 59 | 40764 | 70 | |
XM_017027488.1 | XP_016882977.1 | 59 - 60 | 40830 | 70 | |
XM_017027489.1 | XP_016882978.1 | 56 - 57 | 40676 | 70 | |
XM_017027490.1 | XP_016882979.1 | 54 - 55 | 40365 | 70 | |
XM_017027491.1 | XP_016882980.1 | 54 - 55 | 40365 | 70 | |
XM_017027492.1 | XP_016882981.1 | 54 - 55 | 40362 | 70 | |
XM_017027493.1 | XP_016882982.1 | 54 - 55 | 40362 | 70 | |
XM_017027494.1 | XP_016882983.1 | 54 - 55 | 40362 | 70 | |
XM_017027495.1 | XP_016882984.1 | 54 - 55 | 40362 | 70 | |
XM_017027497.1 | XP_016882986.1 | 34 - 35 | 38487 | 70 | |
XM_017027499.1 | XP_016882988.1 | 59 - 60 | 31617 | 70 | |
XM_017027500.1 | XP_016882989.1 | 58 - 59 | 31536 | 70 | |
XM_017027501.1 | XP_016882990.1 | 38 - 39 | 3544 | 70 | |
GenBank mRNA | AF361486.3 | - | - | 19010 | 70 |
AK128681.1 | - | 15 - 16 | 2129 | 70 | |
AK128856.1 | - | 22 - 23 | 2170 | 70 | |
AK128857.1 | - | - | 3380 | 70 |