Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPNE5
Entrez Gene ID
57699
Gene Name
copine 5
Gene Aliases
COPN5, CPN5
Chromosome Location
Chr.6: 36740773 - 36839444 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001314018.1 NP_001300947.1 8 - 9 758 78
NM_001314019.1 NP_001300948.1 9 - 10 799 78
NM_001314020.1 NP_001300949.1 7 - 8 696 78
NM_020939.1 NP_065990.1 19 - 20 1555 78
XM_005249247.2 XP_005249304.1 20 - 21 1606 78
XM_011514768.1 XP_011513070.1 20 - 21 1606 78
XM_011514769.1 XP_011513071.1 19 - 20 1555 78
XM_011514770.1 XP_011513072.1 21 - 22 1621 78
XM_011514771.2 XP_011513073.1 20 - 21 1946 78
XM_011514773.2 XP_011513075.1 12 - 13 1035 78
XM_011514775.2 XP_011513077.1 8 - 9 749 78
XM_017011139.1 XP_016866628.1 18 - 19 1476 78
GenBank mRNA AB046819.1 - 19 - 20 2113 78
BC053872.1 - 8 - 9 758 78

View Full Product Details