The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020939.1 | NP_065990.1 | 10 - 11 | 807 | 123 |
XM_005249247.2 | XP_005249304.1 | 11 - 12 | 858 | 123 | |
XM_011514768.1 | XP_011513070.1 | 11 - 12 | 858 | 123 | |
XM_011514769.1 | XP_011513071.1 | 10 - 11 | 807 | 123 | |
XM_011514770.1 | XP_011513072.1 | 12 - 13 | 873 | 123 | |
XM_011514771.2 | XP_011513073.1 | 11 - 12 | 1198 | 123 | |
XM_011514772.1 | XP_011513074.1 | 11 - 12 | 858 | 123 | |
XM_011514773.2 | XP_011513075.1 | 3 - 4 | 287 | 123 | |
XM_017011139.1 | XP_016866628.1 | 9 - 10 | 728 | 123 | |
GenBank mRNA | AB046819.1 | - | 10 - 11 | 1365 | 123 |
AI989513.1 | - | 2 - 3 | 123 | ||
DB320492.1 | - | 2 - 3 | 123 |