Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPNE5
Entrez Gene ID
57699
Gene Name
copine 5
Gene Aliases
COPN5, CPN5
Chromosome Location
Chr.6: 36740773 - 36839444 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020939.1 NP_065990.1 13 - 14 978 92
XM_005249247.2 XP_005249304.1 14 - 15 1029 92
XM_011514768.1 XP_011513070.1 14 - 15 1029 92
XM_011514769.1 XP_011513071.1 13 - 14 978 92
XM_011514770.1 XP_011513072.1 15 - 16 1044 92
XM_011514771.2 XP_011513073.1 14 - 15 1369 92
XM_011514772.1 XP_011513074.1 14 - 15 1029 92
XM_011514773.2 XP_011513075.1 6 - 7 458 92
XM_017011139.1 XP_016866628.1 12 - 13 899 92
GenBank mRNA AB046819.1 - 13 - 14 1536 92

View Full Product Details