Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPNE5
Entrez Gene ID
57699
Gene Name
copine 5
Gene Aliases
COPN5, CPN5
Chromosome Location
Chr.6: 36740773 - 36839444 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001314019.1 NP_001300948.1 3 - 4 235 80

View Full Product Details