Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCAPER
Entrez Gene ID
49855
Gene Name
S-phase cyclin A associated protein in the ER
Gene Aliases
MSTP063, ZNF291, Zfp291
Chromosome Location
Chr.15: 76348185 - 76905455 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145923.1 NP_001139395.1 13 - 14 1811 141
NM_020843.2 NP_065894.2 12 - 13 1668 141
XM_005254417.2 XP_005254474.1 13 - 14 1843 141
XM_005254419.2 XP_005254476.1 13 - 14 1825 141
XM_011521650.1 XP_011519952.1 14 - 15 1928 141
XM_011521652.2 XP_011519954.1 12 - 13 1639 141
XM_011521653.2 XP_011519955.1 12 - 13 1723 141
XM_011521654.2 XP_011519956.1 6 - 7 987 141
XM_017022266.1 XP_016877755.1 13 - 14 1848 141
XM_017022267.1 XP_016877756.1 13 - 14 1830 141
XM_017022268.1 XP_016877757.1 13 - 14 1791 141
XM_017022269.1 XP_016877758.1 13 - 14 1823 141
XM_017022270.1 XP_016877759.1 14 - 15 1981 141
XM_017022271.1 XP_016877760.1 13 - 14 1849 141
XM_017022272.1 XP_016877761.1 13 - 14 1843 141
XM_017022273.1 XP_016877762.1 12 - 13 1705 141
XM_017022274.1 XP_016877763.1 13 - 14 1848 141
XM_017022275.1 XP_016877764.1 12 - 13 1680 141
XM_017022276.1 XP_016877765.1 10 - 11 1535 141
XM_017022277.1 XP_016877766.1 12 - 13 1662 141
XM_017022278.1 XP_016877767.1 11 - 12 1544 141
XM_017022279.1 XP_016877768.1 13 - 14 1845 141
XM_017022280.1 XP_016877769.1 13 - 14 1845 141
XM_017022281.1 XP_016877770.1 13 - 14 1845 141
XM_017022282.1 XP_016877771.1 13 - 14 1844 141
XM_017022283.1 XP_016877772.1 13 - 14 1825 141
XM_017022284.1 XP_016877773.1 13 - 14 1856 141
GenBank mRNA AB040887.1 - 12 - 13 1673 141
AF119814.1 - 6 - 7 875 141
AF242528.1 - 12 - 13 1643 141
AK056095.1 - 13 - 14 1832 141
AK056515.1 - 15 - 16 1952 141
BX647285.1 - 13 - 14 1811 141

View Full Product Details