Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCAPER
Entrez Gene ID
49855
Gene Name
S-phase cyclin A associated protein in the ER
Gene Aliases
MSTP063, ZNF291, Zfp291
Chromosome Location
Chr.15: 76348185 - 76905455 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145923.1 NP_001139395.1 28 - 29 3900 71
NM_020843.2 NP_065894.2 27 - 28 3757 71
XM_005254417.2 XP_005254474.1 28 - 29 3932 71
XM_005254419.2 XP_005254476.1 28 - 29 3914 71
XM_011521650.1 XP_011519952.1 29 - 30 4017 71
XM_011521652.2 XP_011519954.1 27 - 28 3728 71
XM_011521653.2 XP_011519955.1 27 - 28 3812 71
XM_011521654.2 XP_011519956.1 21 - 22 3076 71
XM_011521656.2 XP_011519958.1 14 - 15 1933 71
XM_017022266.1 XP_016877755.1 28 - 29 3937 71
XM_017022267.1 XP_016877756.1 28 - 29 3919 71
XM_017022268.1 XP_016877757.1 28 - 29 3880 71
XM_017022269.1 XP_016877758.1 28 - 29 3912 71
XM_017022270.1 XP_016877759.1 29 - 30 4070 71
XM_017022271.1 XP_016877760.1 27 - 28 3782 71
XM_017022272.1 XP_016877761.1 27 - 28 3776 71
XM_017022273.1 XP_016877762.1 27 - 28 3794 71
XM_017022275.1 XP_016877764.1 27 - 28 3769 71
XM_017022276.1 XP_016877765.1 25 - 26 3624 71
XM_017022277.1 XP_016877766.1 27 - 28 3751 71
XM_017022278.1 XP_016877767.1 26 - 27 3633 71
GenBank mRNA AF119814.1 - 21 - 22 2964 71
AF242528.1 - 27 - 28 3732 71
AL137612.1 - 2 - 3 297 71
BC015212.1 - 3 - 4 629 71
BT006762.1 - 2 - 3 374 71
BX647285.1 - 29 - 30 3900 71

View Full Product Details