Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCAPER
Entrez Gene ID
49855
Gene Name
S-phase cyclin A associated protein in the ER
Gene Aliases
MSTP063, ZNF291, Zfp291
Chromosome Location
Chr.15: 76348185 - 76905455 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145923.1 NP_001139395.1 5 - 6 536 77
NM_020843.2 NP_065894.2 5 - 6 552 77
XM_005254417.2 XP_005254474.1 6 - 7 709 77
XM_005254419.2 XP_005254476.1 6 - 7 709 77
XM_011521650.1 XP_011519952.1 7 - 8 794 77
XM_011521652.2 XP_011519954.1 5 - 6 505 77
XM_011521653.2 XP_011519955.1 5 - 6 589 77
XM_017022266.1 XP_016877755.1 6 - 7 714 77
XM_017022267.1 XP_016877756.1 6 - 7 714 77
XM_017022268.1 XP_016877757.1 6 - 7 657 77
XM_017022269.1 XP_016877758.1 6 - 7 707 77
XM_017022270.1 XP_016877759.1 7 - 8 865 77
XM_017022271.1 XP_016877760.1 6 - 7 715 77
XM_017022272.1 XP_016877761.1 6 - 7 709 77
XM_017022273.1 XP_016877762.1 5 - 6 589 77
XM_017022274.1 XP_016877763.1 6 - 7 714 77
XM_017022275.1 XP_016877764.1 6 - 7 707 77
XM_017022276.1 XP_016877765.1 3 - 4 362 77
XM_017022277.1 XP_016877766.1 6 - 7 707 77
XM_017022278.1 XP_016877767.1 5 - 6 589 77
XM_017022279.1 XP_016877768.1 6 - 7 711 77
XM_017022280.1 XP_016877769.1 6 - 7 711 77
XM_017022281.1 XP_016877770.1 6 - 7 711 77
XM_017022282.1 XP_016877771.1 6 - 7 710 77
XM_017022283.1 XP_016877772.1 6 - 7 709 77
XM_017022284.1 XP_016877773.1 6 - 7 722 77
GenBank mRNA AB040887.1 - 5 - 6 539 77
AF242528.1 - 5 - 6 527 77
AK056095.1 - 6 - 7 698 77
AK056515.1 - 8 - 9 836 77
BC070197.1 - 6 - 7 569 77
BC107415.1 - 6 - 7 593 77
BX647285.1 - 5 - 6 536 77

View Full Product Details