Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDR1
Entrez Gene ID
780
Gene Name
discoidin domain receptor tyrosine kinase 1
Gene Aliases
CAK, CD167, DDR, EDDR1, HGK2, MCK10, NEP, NTRK4, PTK3, PTK3A, RTK6, TRKE
Chromosome Location
Chr.6: 30880909 - 30900156 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001202521.1 NP_001189450.1 10 - 11 1555 78
NM_001202522.1 NP_001189451.1 10 - 11 1473 78
GenBank mRNA AF353182.1 - 2 - 3 235 78
AF353183.1 - 2 - 3 153 78

View Full Product Details