The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004882.3 | NP_004873.3 | 2 - 3 | 261 | 152 |
GenBank mRNA | AK292389.1 | - | 2 - 3 | 244 | 152 |
BC015040.1 | - | 2 - 3 | 215 | 152 | |
BC021175.1 | - | 2 - 3 | 229 | 152 | |
BC031018.1 | - | 2 - 3 | 192 | 152 | |
BC038987.1 | - | 2 - 3 | 236 | 152 | |
BC046098.2 | - | 2 - 3 | 212 | 152 | |
BU596189.1 | - | 2 - 3 | 264 | 152 | |
U03644.1 | - | 2 - 3 | 200 | 152 |