The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290149.1 | NP_001277078.1 | 3 - 4 | 771 | 58 |
NM_001290153.1 | NP_001277082.1 | 2 - 3 | 449 | 58 | |
NM_001290154.1 | NP_001277083.1 | 2 - 3 | 449 | 58 | |
NM_199249.2 | NP_954857.1 | 2 - 3 | 449 | 58 | |
GenBank mRNA | BC051842.1 | - | 2 - 3 | 493 | 58 |
BG482545.1 | - | 2 - 3 | 501 | 58 | |
BG746959.1 | - | 1 - 2 | 180 | 58 | |
BG746996.1 | - | 1 - 2 | 181 | 58 | |
BG824338.1 | - | 1 - 2 | 181 | 58 | |
BM802982.1 | - | 2 - 3 | 220 | 58 | |
BQ890364.1 | - | 1 - 2 | 178 | 58 | |
BX508552.1 | - | 2 - 3 | 226 | 58 |