The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205263.1 | NP_001192192.1 | 12 - 13 | 2667 | 89 |
NM_012415.3 | NP_036547.1 | 14 - 15 | 2644 | 89 | |
GenBank mRNA | AF112481.1 | - | 14 - 15 | 2593 | 89 |
AK290437.1 | - | 14 - 15 | 2614 | 89 | |
AL133578.1 | - | 8 - 9 | 1525 | 89 | |
BC001965.2 | - | 14 - 15 | 2586 | 89 |