The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002473.5 | NP_002464.1 | 2 - 3 | 608 | 73 |
XM_011530197.2 | XP_011528499.1 | 3 - 4 | 463 | 73 | |
XM_017028803.1 | XP_016884292.1 | 2 - 3 | 363 | 73 | |
XM_017028804.1 | XP_016884293.1 | 2 - 3 | 363 | 73 | |
XM_017028805.1 | XP_016884294.1 | 2 - 3 | 363 | 73 | |
XM_017028806.1 | XP_016884295.1 | 2 - 3 | 363 | 73 | |
GenBank mRNA | AB191263.1 | - | 2 - 3 | 508 | 73 |
AB290175.1 | - | 2 - 3 | 401 | 73 | |
AK131080.1 | - | 2 - 3 | 474 | 73 | |
AK291609.1 | - | 2 - 3 | 506 | 73 | |
AK304840.1 | - | 2 - 3 | 502 | 73 | |
BC049849.1 | - | 2 - 3 | 477 | 73 | |
BC090921.1 | - | 2 - 3 | 414 | 73 | |
CR456526.1 | - | 2 - 3 | 409 | 73 | |
KU159427.1 | - | - | 541 | 73 | |
M69180.1 | - | 1 - 2 | 347 | 73 | |
M81105.1 | - | 2 - 3 | 467 | 73 |