The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144072.1 | NP_001137544.1 | 2 - 3 | 644 | 86 |
NR_026644.1 | 2 - 3 | 788 | 86 | ||
XM_006719948.3 | XP_006720011.1 | 3 - 4 | 354 | 86 | |
XM_011521084.2 | XP_011519386.1 | 2 - 3 | 209 | 86 | |
GenBank mRNA | AK075419.1 | - | 2 - 3 | 215 | 86 |
AK129861.1 | - | 2 - 3 | 243 | 86 | |
AK290342.1 | - | 2 - 3 | 644 | 86 | |
BC121138.1 | - | 2 - 3 | 387 | 86 | |
BC121139.1 | - | 2 - 3 | 387 | 86 |