Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MFF
Entrez Gene ID
56947
Gene Name
mitochondrial fission factor
Gene Aliases
C2orf33, GL004
Chromosome Location
Chr.2: 227325151 - 227357836 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277061.1 NP_001263990.1 9 - 10 1341 78
NM_001277062.1 NP_001263991.1 7 - 8 1088 78
NM_001277064.1 NP_001263993.1 6 - 7 929 78
NM_001277068.1 NP_001263997.1 6 - 7 694 78
NM_020194.5 NP_064579.3 8 - 9 1078 78
XM_005246682.1 XP_005246739.1 8 - 9 1186 78
XM_005246683.1 XP_005246740.1 8 - 9 1083 78
XM_005246685.3 XP_005246742.1 8 - 9 1102 78
XM_005246687.3 XP_005246744.1 7 - 8 1027 78
XM_006712637.1 XP_006712700.1 7 - 8 985 78
XM_006712638.1 XP_006712701.1 6 - 7 910 78
XM_006712639.2 XP_006712702.1 5 - 6 751 78
XM_011511500.1 XP_011509802.1 9 - 10 1133 78
XM_011511501.1 XP_011509803.1 8 - 9 1154 78
XM_017004512.1 XP_016860001.1 8 - 9 1250 78
GenBank mRNA AF246239.1 - 8 - 9 1078 78
AF258660.1 - 9 - 10 1196 78
AK026137.1 - 7 - 8 989 78
AL517791.3 - 6 - 7 804 78
AL833032.1 - 2 - 3 718 78
BC000797.1 - 5 - 6 601 78
DA737008.1 - 6 - 7 668 78

View Full Product Details