Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MFF
Entrez Gene ID
56947
Gene Name
mitochondrial fission factor
Gene Aliases
C2orf33, GL004
Chromosome Location
Chr.2: 227325151 - 227357836 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277062.1 NP_001263991.1 5 - 6 873 64
NM_001277067.1 NP_001263996.1 5 - 6 619 64
XM_005246682.1 XP_005246739.1 6 - 7 971 64
XM_005246684.1 XP_005246741.1 6 - 7 971 64
XM_006712638.1 XP_006712701.1 4 - 5 695 64
XM_017004512.1 XP_016860001.1 6 - 7 1035 64
XM_017004513.1 XP_016860002.1 5 - 6 793 64
XM_017004514.1 XP_016860003.1 4 - 5 695 64
GenBank mRNA AK026137.1 - 5 - 6 774 64
BQ653061.1 - 3 - 4 344 64

View Full Product Details