The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025243.3 | NP_079519.1 | 6 - 6 | 2294 | 84 |
XM_005246874.3 | XP_005246931.1 | 7 - 7 | 2457 | 84 | |
XM_011511931.2 | XP_011510233.1 | 6 - 6 | 2549 | 84 | |
XM_011511932.1 | XP_011510234.1 | 6 - 6 | 2371 | 84 | |
XM_011511933.1 | XP_011510235.1 | 6 - 6 | 2637 | 84 | |
XM_017005030.1 | XP_016860519.1 | 7 - 7 | 2639 | 84 | |
XM_017005031.1 | XP_016860520.1 | 7 - 7 | 2618 | 84 | |
XM_017005032.1 | XP_016860521.1 | 7 - 7 | 2499 | 84 | |
XM_017005033.1 | XP_016860522.1 | 7 - 7 | 2813 | 84 | |
XM_017005034.1 | XP_016860523.1 | 7 - 7 | 2743 | 84 | |
GenBank mRNA | AF283317.1 | - | 6 - 6 | 2275 | 84 |