The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003852.3 | NP_003843.3 | 18 - 19 | 3056 | 149 |
NM_015905.2 | NP_056989.2 | 18 - 19 | 3158 | 149 | |
GenBank mRNA | AF009353.1 | - | 18 - 19 | 2829 | 149 |
AF119042.1 | - | 18 - 19 | 2957 | 149 | |
AK075306.1 | - | 18 - 19 | 3158 | 149 | |
AK302679.1 | - | 19 - 20 | 3031 | 149 | |
AK312610.1 | - | 18 - 19 | 3184 | 149 | |
BC028689.2 | - | 18 - 19 | 3056 | 149 |