The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135213.1 | NP_001128685.1 | 11 - 12 | 1060 | 61 |
NM_001135214.1 | NP_001128686.1 | 10 - 11 | 1054 | 61 | |
NM_014776.3 | NP_055591.2 | 10 - 11 | 1054 | 61 | |
NM_057169.3 | NP_476510.1 | 10 - 11 | 1054 | 61 | |
NM_057170.3 | NP_476511.1 | 10 - 11 | 1054 | 61 | |
NM_139201.2 | NP_631940.2 | 10 - 11 | 1054 | 61 | |
XM_005253997.3 | XP_005254054.1 | 10 - 11 | 1046 | 61 | |
XM_005254000.3 | XP_005254057.1 | 10 - 11 | 1046 | 61 | |
XM_006719707.3 | XP_006719770.1 | 10 - 11 | 1049 | 61 | |
XM_006719708.3 | XP_006719771.1 | 10 - 11 | 1049 | 61 | |
XM_006719709.3 | XP_006719772.1 | 10 - 11 | 1049 | 61 | |
XM_006719712.3 | XP_006719775.1 | 10 - 11 | 1049 | 61 | |
XM_006719713.3 | XP_006719776.1 | 10 - 11 | 1046 | 61 | |
XM_006719714.3 | XP_006719777.1 | 10 - 11 | 1049 | 61 | |
XM_017020258.1 | XP_016875747.1 | 10 - 11 | 1046 | 61 | |
XM_017020259.1 | XP_016875748.1 | 10 - 11 | 1046 | 61 | |
XM_017020260.1 | XP_016875749.1 | 10 - 11 | 1046 | 61 | |
XM_017020261.1 | XP_016875750.1 | 10 - 11 | 1041 | 61 | |
GenBank mRNA | AB451428.1 | - | 10 - 11 | 889 | 61 |
AF124491.1 | - | 10 - 11 | 889 | 61 | |
AK222736.1 | - | 10 - 11 | 961 | 61 | |
AK303198.1 | - | 9 - 10 | 755 | 61 | |
AK304240.1 | - | 10 - 11 | 1038 | 61 | |
BC001379.2 | - | 10 - 11 | 954 | 61 | |
BC014223.2 | - | 9 - 10 | 805 | 61 | |
BT007312.1 | - | 10 - 11 | 889 | 61 | |
CR533568.1 | - | 10 - 11 | 889 | 61 | |
D63482.2 | - | 11 - 12 | 966 | 61 |