Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ESF1
Entrez Gene ID
51575
Gene Name
ESF1 nucleolar pre-rRNA processing protein homolog
Gene Aliases
ABTAP, C20orf6, HDCMC28P, bA526K24.1
Chromosome Location
Chr.20: 13714322 - 13784933 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276380.1 NP_001263309.1 11 - 12 2134 127
NM_016649.3 NP_057733.2 11 - 12 2145 127
XM_017027874.1 XP_016883363.1 11 - 12 3597 127
GenBank mRNA AK074299.1 - 10 - 11 1631 127

View Full Product Details