The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138777.3 | NP_620132.1 | 2 - 3 | 545 | 59 |
NM_199177.2 | NP_954646.1 | 1 - 2 | 209 | 59 | |
XM_005252302.3 | XP_005252359.1 | 3 - 4 | 613 | 59 | |
XM_011519183.2 | XP_011517485.1 | 3 - 4 | 868 | 59 | |
XM_011519184.2 | XP_011517486.1 | 3 - 4 | 378 | 59 | |
XM_011519185.2 | XP_011517487.1 | 3 - 4 | 613 | 59 | |
XM_011519186.1 | XP_011517488.1 | 2 - 3 | 293 | 59 | |
XM_017015286.1 | XP_016870775.1 | 3 - 4 | 613 | 59 | |
XM_017015287.1 | XP_016870776.1 | 2 - 3 | 293 | 59 | |
GenBank mRNA | AK291603.1 | - | 2 - 3 | 235 | 59 |
AK291809.1 | - | 2 - 3 | 274 | 59 | |
AK292790.1 | - | 2 - 3 | 274 | 59 | |
AK298046.1 | - | 2 - 3 | 264 | 59 | |
AK316266.1 | - | 3 - 4 | 562 | 59 | |
BC013049.1 | - | 1 - 2 | 206 | 59 | |
BC053604.1 | - | 1 - 2 | 207 | 59 | |
BF982227.1 | - | - | 280 | 59 |